Orders LinkContactUsLinkComponent
Sulphite standard solution
Sulphite standard solution
Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST), 5000 mg/l as SO3, 16
Codice catalogo HACH2267410
Nome del fornitore:   Hach
Sulphite standard solution
Codice catalogo HACH2267410
Nome del fornitore:  Hach
Numero CAS:  
Sulphite standard solution
Sulphite standard solution
Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST), 5000 mg/l as SO3, 16
Numero CAS:  
Price
118.00 SFr.
Log in per disponibilità
1 * 16 ST
Product Details & Documents
This equivalent standard is prepared with stabilised sodium thiosulfate, since sulphite is not a stable in solution.
Documents
Specifiche
Specifiche
  • Concentrazione:
    5000 mg/l as SO<sub>3</sub>
  • Conf:
    16
  • Descrizione:
    Sulfite standard solution, pack of 16×10 ml voluette ampoules (NIST)