Hydrophobic interaction chromatography columns, TSK-GEL®
Guardgel kit, for P/N 07573, Precolonna, Phenyl-5PW, 20 µm
Codice catalogo TOYO0007652
Nome del fornitore: TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Codice catalogo TOYO0007652
Nome del fornitore: TOSOH Bioscience
Vai a
Product Details & Documents
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
- Rigid polymeric base resin
- Eliminates undesirable interactions with column hardware
- Wide pH range (2 to 12) enables robust cleaning options
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Formato della colonna:Precolonna
- Dimensioni particelle:20 µm
- Descrizione:Guardgel kit, for P/N 07573
- Fase:Phenyl-5PW