Cation exchange chromatography columns, TSK-GEL® CM-3SW
Guardgel kit, Precolonna, CM-SW, -, 20 µm
Codice catalogo TOYO0007650
Nome del fornitore: TOSOH Bioscience
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Codice catalogo TOYO0007650
Nome del fornitore: TOSOH Bioscience
Vai a
Product Details & Documents
TSK-GEL® CM-3SW are silica based stainless steel columns packed with spherical silica beads which are modified with carboxymethyl (CM) ligands to provide a weak cation exchange column. Designed for the separation and analysis of proteins, peptides, and other biologically active molecules.
Specifiche
- Formato della colonna:Precolonna
- Dimensioni particelle:20 µm
- Dimensioni pori:-
- Descrizione:Guardgel kit
- Seq:03
- Fase:CM-SW