Orders LinkContactUsLinkComponent
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Guardgel kit, Precolonna, CM-SW, -, 20 µm
Codice catalogo TOYO0007650
Nome del fornitore:   TOSOH Bioscience
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Codice catalogo TOYO0007650
Nome del fornitore:  TOSOH Bioscience
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Guardgel kit, Precolonna, CM-SW, -, 20 µm
Price
SFr. 973.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
TSK-GEL® CM-3SW are silica based stainless steel columns packed with spherical silica beads which are modified with carboxymethyl (CM) ligands to provide a weak cation exchange column. Designed for the separation and analysis of proteins, peptides, and other biologically active molecules.
Specifiche
  • Formato della colonna:
    Precolonna
  • Dimensioni particelle:
    20 µm
  • Dimensioni pori:
    -
  • Descrizione:
    Guardgel kit
  • Seq:
    03
  • Fase:
    CM-SW