Cation exchange chromatography columns, TSK-GEL® CM-3SW
Cation exchange chromatography column, CM-2SW, Analitica, CM-2SW, Acciaio inossidabile, 125 Å, 5 µm, 4,60 mm, 250 mm, Silica
Codice catalogo TOYO0007167
Nome del fornitore: TOSOH Bioscience
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Codice catalogo TOYO0007167
Nome del fornitore: TOSOH Bioscience
Vai a
Product Details & Documents
TSK-GEL® CM-3SW are silica based stainless steel columns packed with spherical silica beads which are modified with carboxymethyl (CM) ligands to provide a weak cation exchange column. Designed for the separation and analysis of proteins, peptides, and other biologically active molecules.
Specifiche
- Formato della colonna:Analitica
- Ø int.:4,60 mm
- Dimensioni particelle:5 µm
- Dimensioni pori:125 Å
- Descrizione:Cation exchange chromatography column, CM-2SW
- Lunghezza:250 mm
- Seq:01
- Colonna:Acciaio inossidabile
- Fase:CM-2SW
- Materiale d'imballaggio:Silica
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



