Capillary columns, CP-Wax™
on 7" cage, Capillare, CP-Wax 52 CB, 1,00 µm, 0,53 mm, 10 m
Codice catalogo 558-0032
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
The CP-Wax™ lines of chemically bonded columns offer a broad range of selectivities for method optimisation. The backbone of this series stationary phases is made of a polyethylene glycol.
Specifiche
- Formato della colonna:Capillare
- Spessore dello strato:1,00 µm
- Ø int.:0,53 mm
- Descrizione:on 7" cage
- Lunghezza:10 m
- Seq:33
- Fase:CP-Wax 52 CB