Orders LinkContactUsLinkComponent
Salt bridges
Replacement salt bridge, AL110, double liquid junction, ceramic junction, 250 mm length, for Ø 7,5 or 8 mm reference electrodes
Codice catalogo HACHB40A550
Nome del fornitore:   Hach
Salt bridges
Codice catalogo HACHB40A550
Nome del fornitore:  Hach
Salt bridges
Salt bridges
Replacement salt bridge, AL110, double liquid junction, ceramic junction, 250 mm length, for Ø 7,5 or 8 mm reference electrodes
Price
SFr. 116.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.
Specifiche
  • Descrizione:
    Replacement salt bridge, AL110, double liquid junction, ceramic junction, 250 mm length, for Ø 7,5 or 8 mm reference electrodes
  • Seq:
    02