Orders LinkContactUsLinkComponent
Salt bridges
Salt bridges
Replacement salt bridge, AL120, double liquid junction, ceramic junction, 138 mm length, for reference electrodes with a ground joint
Codice catalogo HACHB40A610
Nome del fornitore:   Hach
Salt bridges
Codice catalogo HACHB40A610
Nome del fornitore:  Hach
Salt bridges
Salt bridges
Replacement salt bridge, AL120, double liquid junction, ceramic junction, 138 mm length, for reference electrodes with a ground joint
Price
422.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.
Specifiche
  • Descrizione:
    Replacement salt bridge, AL120, double liquid junction, ceramic junction, 138 mm length, for reference electrodes with a ground joint
  • Seq:
    03