Digital Solutions
Applications
Support
About
Switzerland
Orders Link
ContactUsLinkComponent
Vassoi/Piatti/Contenitori
Vassoi per strumenti
Instrument trays
Instrument trays
200×175×15 mm
BOCH8480
:
BOCHEM
Instrument trays
BOCH8480
:
BOCHEM
Instrument trays
200×175×15 mm
SFr. 82.70
1 * 1 ST
-
+
Made of 18/10 steel.
Very good chemical resistance
Very good temperature resistance
Sturdy
L×P×A:
200×175×15 mm
Vassoi da laboratorio
#: 216-2101
: KARTELL