Pinze per burette
Steel, zinc plated, fingers with silicone coating, 0 - 20 mm, Una buretta
Codice catalogo BOCH5476
Nome del fornitore: BOCHEM
Vai a
Product Details & Documents
Burette clamps made of zinc plated stainless steel.
- Fingers are covered with silicone coating to protect clamped items
- Single sided or double sided available
Specifiche
- Per:Una buretta
- Larghezza di serraggio:0 - 20 mm
- Materiale:Steel, zinc plated, fingers with silicone coating