Gel casting system, MultiCaster
UVT tray for MultiCaster
Codice catalogo 730-0360
Nome del fornitore: EMBITEC
Vai a
Product Details & Documents
Moulded PC. Compatible with the RunOne™ electrophoresis system.
- Simple to use - no gaskets, clamps or tape required
- Flexible with options for well size, well configuration, number of wells and resolving area
Specifiche
- Descrizione:UVT tray for MultiCaster
- Seq:05
- Cat. No.:730-0360
- Dimensioni gel:—