Etichette per vetrini
Etichette 11×22 m in fogli, stampabili, Paper, nero su bianco, 11×22 mm
Catalog # 630-1838
Supplier: PRECISION DYNAMICS TIMEM
Jump to:
Product Details & Documents
These thin labels are suitable for use with microscopy slides.
- Permanent adhesive
- No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Specifications
- Colore:nero su bianco
- Descrizione:Etichette 11×22 m in fogli, stampabili
- Dimensioni:11×22 mm
- Materiale:Paper
- Seq:002