Potassium chloride standard solution
Potassium chloride standard solution, 35,0 ppt salinity, 53,0 mS/cm, 500 mL
Supplier: Hach
Jump to:
Product Details & Documents
For calibrations/accuracy checks on conductivity/salinity meters.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Conf:500 mL
- Descrizione:Potassium chloride standard solution, 35,0 ppt salinity
- Conduttanza:53,0 mS/cm