Orders LinkContactUsLinkComponent
Potassium chloride standard solution
Potassium chloride standard solution
Potassium chloride standard solution, 35,0 ppt salinity, 53,0 mS/cm, 500 mL
Catalog # HACH2714349
Supplier:   Hach
Potassium chloride standard solution
Catalog # HACH2714349
Supplier:  Hach
CAS Number:  
Potassium chloride standard solution
Potassium chloride standard solution
Potassium chloride standard solution, 35,0 ppt salinity, 53,0 mS/cm, 500 mL
CAS Number:  
Price
SFr. 131.00
Log in for availability
1 * 500 mL
Product Details & Documents
For calibrations/accuracy checks on conductivity/salinity meters.
Documents
Specifications
Specifications
  • Conf:
    500 mL
  • Descrizione:
    Potassium chloride standard solution, 35,0 ppt salinity
  • Conduttanza:
    53,0 mS/cm