Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Non-Flexible Needles

Non-Flexible Needles

Curved, with 2,25 mm stainless steel ball tip, 18, 38 mm, Stainless steel ball
Codice catalogo POPR7912
Nome del fornitore:   CADENCE SCIENCE
Non-Flexible Needles
Codice catalogo POPR7912
Nome del fornitore:  CADENCE SCIENCE
Non-Flexible Needles
Non-Flexible Needles

Curved, with 2,25 mm stainless steel ball tip, 18, 38 mm, Stainless steel ball

Price
SFr. 3’810.00
Log in per disponibilità
1 * 12 ST
Product Details & Documents

All needles are made with type 304 stainless steel cannulae. These all-metal, reusable needles are designed for oral gavage procedures in laboratory animals. The precision assembly of stainless steel balls and 304 stainless steel tubing ensures a quality needle suited for feeding and dosing without causing trauma to the mouth or esophagus. Ideal for pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals.

  • Fixed shape from stainless steel tubing
  • With stainless steel ball tips
  • Can be cleaned and re-used as needed
Specifiche
  • Gauge:
    18
  • Tipo punta:
    Stainless steel ball
  • Descrizione:
    Curved, with 2,25 mm stainless steel ball tip
  • Lunghezza:
    38 mm
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..