Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Non-Flexible Needles

Curved, with 2,25 mm stainless steel ball tip, 20, 38 mm, Stainless steel ball
Codice catalogo POPR7910
Nome del fornitore:   CADENCE SCIENCE
Non-Flexible Needles
Codice catalogo POPR7910
Nome del fornitore:  CADENCE SCIENCE
Non-Flexible Needles
Non-Flexible Needles

Curved, with 2,25 mm stainless steel ball tip, 20, 38 mm, Stainless steel ball

Price
SFr. 1’710.00
Log in per disponibilità
1 * 12 ST
Product Details & Documents

All needles are made with type 304 stainless steel cannulae. These all-metal, reusable needles are designed for oral gavage procedures in laboratory animals. The precision assembly of stainless steel balls and 304 stainless steel tubing ensures a quality needle suited for feeding and dosing without causing trauma to the mouth or esophagus. Ideal for pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals.

  • Fixed shape from stainless steel tubing
  • With stainless steel ball tips
  • Can be cleaned and re-used as needed
Specifiche
  • Gauge:
    20
  • Tipo punta:
    Stainless steel ball
  • Descrizione:
    Curved, with 2,25 mm stainless steel ball tip
  • Lunghezza:
    38 mm
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.