Immunology tubes
4 ml, Fondo sferico, Microlon 600
Codice catalogo 738-0001
Nome del fornitore: Greiner Bio-One
Immunology tubes
Codice catalogo 738-0001
Nome del fornitore: Greiner Bio-One
Vai a
Product Details & Documents
PS, clear. Star bottomed tubes have an increased surface area and thus make it possible to bind larger amounts of antigens or antibodies.
Immuno tubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone).
Available with either MICROLON® 200 (medium binding) and MICROLON® 600 (high binding) surfaces.
Ø×L: 12×75 mm
Specifiche
- Capacità:4 ml
- Descrizione:Fondo sferico, Microlon 600
- Seq:01
- Cat. No.:738-0001
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:ISO 14001 (241002); ISO 50001 (241002)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



