Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Soluzioni ISA

Soluzioni ISA

Complextion ionic strength adjuster (CISA) for chloride electrode, 1 L, Ion selective electrode reagents, Bottle, 1
Codice catalogo 662-0838
Nome del fornitore:   Thermo Orion
Soluzioni ISA
Codice catalogo 662-0838
Nome del fornitore:  Thermo Orion
Numero CAS:  
Soluzioni ISA
Soluzioni ISA

Complextion ionic strength adjuster (CISA) for chloride electrode, 1 L, Ion selective electrode reagents, Bottle, 1

Numero CAS:  
Price
SFr. 425.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

  • Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
  • Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
  • Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
  • Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences

Concentration : 10 ppm

For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes

Specifiche

Specifiche

  • Tipo di conf.:
    Bottle
  • Conf:
    1
  • Descrizione:
    Complextion ionic strength adjuster (CISA) for chloride electrode, 1 L
  • Tipo:
    Ion selective electrode reagents

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.