2-Propanolo ≥99.0%, GPR RECTAPUR®
bottiglia in plastica, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0,786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambiente
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Documents
Certificates
Search by Lot Number14510279.pdf P20_VWRC20839.297_SPEC_EN.pdf P20_VWRC20839.366_SPEC_EN.pdf P20_VWRC20839.468_SPEC_EN.pdf P20_VWRC20839.550_SPEC_EN.pdf
Product specification
More
1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Acidity:Passes test
- Acidity or alkalinity:Passes test
- Benzene and related substances:Passes test
- IR Spectrum:Passes test
- Peroxides:Passes test
- Boiling point:82.2 - 82.6 °C
- Density (20/20):0.785 - 0.789
- Density (25/25):0.783 - 0.790
- Indice di rifrazione (20°C):1.376 - 1.378
- Evaporation residue:Max. 20 ppm
- Water:Max. 0.5 %
Specifications
- Tipo di conf.:bottiglia in plastica
- Conf:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



