Alcol iso butilico GPR RECTAPUR Fusto in metallo 25L 1 * 25 L
20836.468
Frequently Bought Together
Trietilammina ≥99.6%, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
N,N-Dimetilformammide ≥99.9%, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acqua, AnalaR NORMAPUR® ISO 3696 Grade 3 per analisi
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
: VWR Chemicals