Orders LinkContactUsLinkComponent
Tubidity standards for 2100A
Tubidity standards for 2100A
StabCal® standard calibration kit for model 2100A, 1,0 NTU, Sealed vials, 4× sealed vial, 1 KIT
Codice catalogo HACH2659105
Nome del fornitore:   Hach
Tubidity standards for 2100A
Codice catalogo HACH2659105
Nome del fornitore:  Hach
Numero CAS:  
Tubidity standards for 2100A
Tubidity standards for 2100A
StabCal® standard calibration kit for model 2100A, 1,0 NTU, Sealed vials, 4× sealed vial, 1 KIT
Numero CAS:  
Price
SFr. 776.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazin solutions
  • Based on Formazin Primary Standards
  • Enhanced stability saves time and labour
  • Confidence in results
Documents
Specifiche
Specifiche
  • Tipo di conf.:
    Sealed vials
  • Conf:
    1 KIT
  • Torbidità:
    1,0 NTU
  • Descrizione:
    StabCal® standard calibration kit for model 2100A
  • Pk Info:
    4× sealed vial