Test kits, digital titration method
Chlorine (Free and Total) reagent set, high range, 0 - 3 mg/l, 100 Tests
Codice catalogo HACH2445300
Nome del fornitore: Hach
Test kits, digital titration method
Codice catalogo HACH2445300
Nome del fornitore: Hach
Numero CAS:
Vai a
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
- Reagents are pre-prepared
- Exact comparison of colour
- Rugged carrying case for analysis in the field
: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifiche
Specifiche
- Conf:100 Tests
- Descrizione:Chlorine (Free and Total) reagent set, high range
- Range di misura:0 - 3 mg/l