Multiparameter test kits, model HA-62
For hardness (as CaCO₃), iron and pH, HA-62B, Hardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 10, 1 KIT
Codice catalogo HACH183702
Nome del fornitore: Hach
Multiparameter test kits, model HA-62
Codice catalogo HACH183702
Nome del fornitore: Hach
Numero CAS:
Vai a
Product Details & Documents
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
- Popular tests in one rugged kit
- Uses color disc and drop count titration methods
Specifiche
Specifiche
- Conf:1 KIT
- Descrizione:For hardness (as CaCO₃), iron and pH, HA-62B
- Range di misura:Hardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 10