Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ AccuCAT

Solid phase extraction cartridges, Bond Elut™ AccuCAT

Straight barrel cartridge, AccuCAT, 40 µm, 6 ml, 200 mg
Codice catalogo VARI12282004
Nome del fornitore:   VARIAN
Solid phase extraction cartridges, Bond Elut™ AccuCAT
Codice catalogo VARI12282004
Nome del fornitore:  VARIAN
Solid phase extraction cartridges, Bond Elut™ AccuCAT
Solid phase extraction cartridges, Bond Elut™ AccuCAT

Straight barrel cartridge, AccuCAT, 40 µm, 6 ml, 200 mg

Price
Richiedi prezzo
Log in per disponibilità
1 * 30 ST
Product Details & Documents

Bond Elut AccuCAT cartridges are mixed bed SPE cartridges consisting of a strong cation exchange (SCX) and a strong anion exchange (SAX) sorbent packed into one bed. AccuCAT is effective for the extraction of acidic, basic and neutral analytes from urine and other biological samples. AccuCAT is particularly effective for catecholamine extraction from bio fluids.

  • Ultra clean, mixed sorbent bed delivers for reproducible extractions
  • SCX and SAX functionalities offer broad analyte extraction potential
  • Compatible with many biological fluids for easy method transfer
Specifiche
  • Dimensioni particelle:
    40 µm
  • Capacità:
    6 ml
  • Descrizione:
    Straight barrel cartridge
  • Seq:
    05
  • Peso letto:
    200 mg
  • Fase:
    AccuCAT
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.