Orders LinkContactUsLinkComponent
Test kits, digital titration method
Test kits, digital titration method
EDTA digital titrator cartridge, 0,0716 M, 1
Catalog # HACH2081701
Supplier:   Hach
Test kits, digital titration method
Catalog # HACH2081701
Supplier:  Hach
CAS Number:  
Test kits, digital titration method
Test kits, digital titration method
EDTA digital titrator cartridge, 0,0716 M, 1
CAS Number:  
Price
SFr. 64.60
Log in for availability
1 * 1 ST
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Conf:
    1
  • Descrizione:
    EDTA digital titrator cartridge, 0,0716 M