Microtest manifolds
Microtest manifold, SS/PTFE, straight, 4-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
Codice catalogo DRUM3-000-090
Nome del fornitore: DRUMMOND
Vai a
Product Details & Documents
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
- Autoclavable
- Use for both sample delivery and aspiration operations
- Supplied complete with cleaning wire for easy cleaning
- Positive Luer lock connection
Specifiche
- Descrizione:Microtest manifold, SS/PTFE, straight, 4-place for 96 well plate, 9 mm centre-to-centre, pin length 11 mm
- Seq:01