Orders LinkContactUsLinkComponent
Distillation apparatus set, general purpose
Distillation apparatus set, general purpose
Cyanide set
Codice catalogo HACH2265800
Nome del fornitore:   Hach
Distillation apparatus set, general purpose
Codice catalogo HACH2265800
Nome del fornitore:  Hach
Distillation apparatus set, general purpose
Distillation apparatus set, general purpose
Cyanide set
Price
885.00 SFr.
Log in per disponibilità
1 * 1 SET
Product Details & Documents
Fast set-up distillation apparatus for sample pretreatment. Distillation sets include only EPAapproved glassware.
  • Easy to set up, only required parts are delivered
  • Illustrated, step-by-step procedures simplify distillation
Adapts to a variety of test needs including all wastewater and water methods requiring sample pretreatment by distillation. Applications include fluoride, ammonia-nitrogen, phenols, selenium, volatile acids and albuminoid nitrogen.
Informazioni per l'ordine: Includes distillation flask, J-tube, condenser, drip tube, erlenmeyer flask, and non-Hg thermometer. Arsenic and cyanide sets are available, these are used together with the general purpose distillation set.
Attenzione: Heater and support apparatus, sold separately, is required for distillation.
Specifiche
  • Descrizione:
    Cyanide set