Sample cuvettes
Sample cuvettes, matched pair, 10/25 ml marked, 25 ml, 1" / 25 mm, Quarzo
Codice catalogo 705-0171
Nome del fornitore: Hach
Vai a
Product Details & Documents
Sample cuvettes for photometric measurements.
- Available with or without lid
- Available reusable (made from glass) or for single use (made from plastics)
Specifiche
- Cammino ottico:1" / 25 mm
- Volume:25 ml
- Materiale finestra:Quarzo
- Descrizione:Sample cuvettes, matched pair, 10/25 ml marked
- Seq:11