Test tubes, with screw cap
50 ml, 115 mm
Codice catalogo KIMA17174
Nome del fornitore: KIMA SAS DI CHIARIN RENZO
Vai a
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
Universal tubes for a wide range of applications.
Specifiche
- Capacità:50 ml
- Lunghezza:115 mm