Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Ion selective electrodes, bromide, Orion™

Ion selective electrodes, bromide, Orion™

Bromide solid-state half-cell, 9435BN, 5×10-6 - 1,0 M (0,40 - 79900 ppm), 0...+80 °C, 9435BN, BNC, 1 m cable
Codice catalogo 662-0842
Nome del fornitore:   Thermo Orion
Ion selective electrodes, bromide, Orion™
Codice catalogo 662-0842
Nome del fornitore:  Thermo Orion
Ion selective electrodes, bromide, Orion™
Ion selective electrodes, bromide, Orion™

Bromide solid-state half-cell, 9435BN, 5×10-6 - 1,0 M (0,40 - 79900 ppm), 0...+80 °C, 9435BN, BNC, 1 m cable

Price
SFr. 1’170.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These electrodes are designed for measuring bromide ions in aqueous solutions quickly, simply and economically.

  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode (9635BNWP) does not require a separate reference electrode, making it convenient to use with small sample volumes

Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately. Screw cap electrode is supplied without cable.

Specifiche
  • Concentrazione:
    5×10-6 - 1,0 M (0,40 - 79900 ppm)
  • Connettore:
    BNC, 1 m cable
  • Tipo:
    9435BN
  • Descrizione:
    Bromide solid-state half-cell, 9435BN
  • Seq:
    02
  • Temperatura:
    0...+80 °C
Accessories

Informazioni sul prodotto

  • Codice catalogo
    Concentrazione
    Conf
    Disponibilità
    Price
  • Bromuro, soluzione standard
    Bromuro, soluzione standard
    ORIO943506
    0,1M
    475 mL
    Log in per disponibilità
    1 * 475 mL - SFr. 136.00

Informazioni sul prodotto

  • Codice catalogo
    Tipo
    Tipo di conf.
    Disponibilità
    Price
  • Regolatore di forza ionica per elettrodo ione-selettivo per cloruro, in flacone
    Regolatore di forza ionica per elettrodo ione-selettivo per cloruro, in flacone
    662-0824
    Ion selective electrode reagents
    Bottle
    Log in per disponibilità
    1 * 475 mL - SFr. 126.00

Informazioni sul prodotto

  • Codice catalogo
    Conf
    Disponibilità
    Price
  • Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    662-1524
    5
    Log in per disponibilità
    1 * 5 ST - SFr. 150.00

Informazioni sul prodotto

  • Codice catalogo
    Conf
    Disponibilità
    Price
  • Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    662-0750
    5
    Log in per disponibilità
    1 * 5 ST - SFr. 150.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.