Vacuum filtering flasks, Büchner, with screwthread connector
2000 ml, 260 mm, 166 mm, 60 mm
Catalog # 511-0777
Supplier: SciLabware
Vacuum filtering flasks, Büchner, with screwthread connector
Catalog # 511-0777
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Side arm features a PTFE screwthread connector for easy and safe fitting of 9 mm bore vacuum tubing
- With tooled neck to accept a rubber stopper or cone
Büchner flasks with heavy wall construction for vacuum filtration applications.
Certifications: Complies with ISO 6556
Specifications
- Ø est:166 mm
- Capacità:2000 ml
- Altezza:260 mm
- Seq:08
- Ø interno collo:60 mm