Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Rotary evaporating flasks

Rotary evaporating flasks

500 ml, 29/32 NS, 160 mm
Codice catalogo 201-8406
Nome del fornitore:   SciLabware
Rotary evaporating flasks
Codice catalogo 201-8406
Nome del fornitore:  SciLabware
Rotary evaporating flasks
Rotary evaporating flasks

500 ml, 29/32 NS, 160 mm

Price
SFr. 84.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents

PYREX®, borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.

Specifiche
  • Cono femmina:
    29/32 NS
  • Capacità:
    500 ml
  • Seq:
    07
  • Altezza totale:
    160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.