Instrument boxes
giallo, 100×215×50 mm
AGEN8036J
Aluminium, anodised.
- Smooth surfaces for fast, easy cleaning
- Coloured
- Good chemical resistance, very good thermal resistance
- Colore:giallo
- Seq:07
- L×P×A:100×215×50 mm