Spatulas
Spatula, analysis type (A), 230 mm
Nome del fornitore: Agencinox
Vai a
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
- Excellent chemical and thermal resistance
- Autoclavable
Documents
Specifiche
- Descrizione:Spatula, analysis type (A)
- Lunghezza:230 mm