Sample bottles for push-on caps
18 ml, 38,5 mm, 30 mm
Codice catalogo 215-2633
Nome del fornitore: GRAVIS
Vai a
Product Details & Documents
Chemistry glass, white or amber, without push-on cap.
- Amber glass to protect light-sensitive materials.
Ordering information: Push-on PE caps are available separately.
Documents
SPVAE00943_TDS_EN
Datasheet
Specifiche
- Capacità:18 ml
- Altezza:38,5 mm
- Ø collo:30 mm