Trypsinising flasks, with screw caps, WHEATON®
300 ml, Top screw cap 38-430
Codice catalogo WHEA355445
Nome del fornitore: DWK Life Sciences
Trypsinising flasks, with screw caps, WHEATON®
Codice catalogo WHEA355445
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Certifications: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Specifiche
- Capacità:300 ml
- Descrizione:Top screw cap 38-430