Tween® 20 (Polysorbate)
bottiglia in vetro, 1 L
Catalog # NADIEC-607-1L
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Tween® 20 (Polysorbate)
Catalog # NADIEC-607-1L
Supplier: NATIONAL DIAGNOSTICS
Marketsource
CAS Number: 9005-64-5
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1,1 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:500-018-3
Specifications
Specifications
- Tipo di conf.:bottiglia in vetro
- Conf:1 L