Ammonio persolfato (APS)
25 g
Catalog # NADIEC-504-25G
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Ammonio persolfato (APS)
Catalog # NADIEC-504-25G
Supplier: NATIONAL DIAGNOSTICS
Marketsource
CAS Number: 7727-54-0
Jump to:
Product Details & Documents
Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate
- CAS Number:7727-54-0
- Formula:(NH₄)₂S₂O₈
- MDL Number:MFCD00003390
- Molecular Weight:228.2 g/mol
- Merck Index:13,00545
- Melting Pt:120 °C
- Density:1,98 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:231-786-5
- UN:1444
- ADR:5.1,III
Specifications
Specifications
- Conf:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



