Mini molecular models: DNA
Mini molecular model of DNA, 11 layers
Catalog # 763-0453
Marketsource
Supplier: SPIRING ENTERPRISES
Jump to:
Product Details & Documents
Mini molecular models of DNA with nucleic acids, five-sided sugar molecules and pyramid-shaped phosphate molecules.
Only correct base pairs can be connected.
Ordering information: Components include colour-coded bases: G, C, A, T.
Specifications
- Descrizione:Mini molecular model of DNA, 11 layers