Staining racks
Tray, PVC, 380×170×80 mm
Katalognr. 631-0337
Anbieter: GLASWARENFABRIK KARL HECHT
Direkt zu
Product Details & Documents
Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or dishes of 380 to 620 mm in length.
Technische Daten
- Descrizione:Tray, PVC
- L×P×A:380×170×80 mm