Infusion bottles
With thread, 300 ml, GL 38
Supplier: APG Europe
Jump to:
Product Details & Documents
Made of type 1 clear glass.
Graduated, with crimp neck or thread, without screw cap.
Documents
Certificates
Search by Lot NumberSpecifications
- Filettatura:GL 38
- Capacità:300 ml
- Descrizione:With thread
- Seq:06