Apparatus - Antonacopoulos
Spare distilling link with 2 cones, 29/32 NS, - NS
Codice catalogo LENZ05434004
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®, Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Dimensione cono:29/32 NS
- Cono femmina:- NS
- Descrizione:Spare distilling link with 2 cones
- Seq:04