/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Il tuo profilo
Cart
Switzerland
Order Entry
Order Status
Il tuo profilo
Cart
Orders Link
ContactUsLinkComponent
Home
Ghiere
Tappi per bottiglie
VWR®, Screw Caps
Print
VWR®, Screw Caps
18 mm, 50/100 ml
Codice catalogo 215-5643
Nome del fornitore:
VWR Collection
VWR®, Screw Caps
Codice catalogo 215-5643
Nome del fornitore:
VWR Collection
Return to Top
VWR®, Screw Caps
18 mm, 50/100 ml
Specifiche
Vai a
Specifiche
Price
SFr. 4.30
Log in
per disponibilità
1 * 10 ST
-
+
famiglie di prodotti
Return Policy
Product Details & Documents
CoC VWR Bottles, Narrow Neck, with Screw Cap
CoC_VWR_ SCREW_CAPS_FOR_NARROW_MOUTH_BOTTLES_WITH_THREAD
VWR® for sampling and sample transportation
VWR® Lab bric-a-brac
Specifiche
Filettatura:
18 mm
Seq:
07
Per bottiglie:
50/100 ml
VWR®, Spray Caps for Narrow Neck Bottles
#: 215-6312
Supplier: VWR Collection
VWR®, Dropper Closures for Narrow Neck Bottles
#: 215-2195
Supplier: VWR Collection
VWR®, bottiglie, collo stretto, rotonde, LDPE, con tappi a vite
#: 215-5638
Supplier: VWR Collection