VISOCOLOR® Colorimetric Test, Chloride
VISOCOLOR® HE Chloride CL 500, Reagents for 300 tests, titration syringe, test tube with ring mark and 2 plastic dropping tips, 5 - 500 mg/L (Cl⁻) - (1 syringe filling), Mercurimetric titration, Visual, 170×125×45 mm (Including packaging), Sea water compatible, 300, 1 KIT
Nome del fornitore: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Chloride
Codice catalogo MANA915004
Nome del fornitore: MACHEREY-NAGEL
Numero CAS:
Vai a
Product Details & Documents
Reagents for the determination of chloride in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifiche
Specifiche
- Metodo chimico:Mercurimetric titration
- Conf:1 KIT
- Descrizione:VISOCOLOR® HE Chloride CL 500
- Dimensioni:170×125×45 mm (Including packaging)
- Tipo:Sea water compatible
- Per strumento:Visual
- Test per kit:300
- Range di misura:5 - 500 mg/L (Cl⁻) - (1 syringe filling)
- Indice:Reagents for 300 tests, titration syringe, test tube with ring mark and 2 plastic dropping tips
- Cat. No.:MANA915004
- Peso:304,5 g (Including packaging)
- Modello:MANA915004
- Piattaforma:Titrimetric test kits
- Durata a magazzino (mesi):24
- Temperatura di conservazione:15...25 °C



