Orders LinkContactUsLinkComponent
Distillatori
Distillatori
160 mm, 14/23 NS, 14/23 NS
  201-0899
Distillatori
  201-0899
Distillatori
Distillatori
160 mm, 14/23 NS, 14/23 NS
SFr. 163.00
1 * 1 ST
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Bridge includes fused Liebig condenser and curved vacuum adapter. NS 14/23 socket for distillation thermometer with 55 mm fitted length. Water and vacuum connection via hose connectors with 8 mm external diameter or water connection with thread GL 14 and vacuum receiver adapter with thread GL 18.
  • Dimensione cono:
    14/23 NS
  • Lunghezza mantello:
    160 mm
  • Cono femmina:
    14/23 NS