Distillatori
160 mm, 14/23 NS, 14/23 NS
201-0899
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Bridge includes fused Liebig condenser and curved vacuum adapter. NS 14/23 socket for distillation thermometer with 55 mm fitted length. Water and vacuum connection via hose connectors with 8 mm external diameter or water connection with thread GL 14 and vacuum receiver adapter with thread GL 18.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Schott tubing
Datasheet
- Dimensione cono:14/23 NS
- Lunghezza mantello:160 mm
- Cono femmina:14/23 NS