BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H)number sequence: 01-24, 300 ml
Codice catalogo WHEA227497-01
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specifiche
- Capacità:300 ml
- Descrizione:BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H)number sequence: 01-24