Heating tapes, HBS
Heating tape HBS, 1,5 m length, 350 W
Codice catalogo HORS020303
Nome del fornitore: HORST
Vai a
Product Details & Documents
The heating tapes are smooth, flexible and have a multiple glass-yarn insulation as well as an additional protection braid around the heating conductors.
Specifiche
- Descrizione:Heating tape HBS, 1,5 m length, 350 W
- Seq:02