Orders LinkContactUsLinkComponent
Control valves, BOLA
Control valves, BOLA
Control valve with thread M 18×2, for tubing outer Ø 8 mm, PTFE
Codice catalogo BOHLE672-54
Nome del fornitore:   Bohlender
Control valves, BOLA
Codice catalogo BOHLE672-54
Nome del fornitore:  Bohlender
Control valves, BOLA
Control valves, BOLA
Control valve with thread M 18×2, for tubing outer Ø 8 mm, PTFE
Price
608.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Body made of PTFE with 2 connections and nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Volume flow can be regulated manually
  • Suitable for panel mounting
  • Easy disassembly for cleaning
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Specifiche
  • Descrizione:
    Control valve with thread M 18×2, for tubing outer Ø 8 mm, PTFE