Etichette per vetrini
Slide labels on sheets, custom imprint, tag, Paper, nero su bianco, 22×22 mm
Codice catalogo 631-9498
Nome del fornitore: PRECISION DYNAMICS TIMEM
Vai a
Product Details & Documents
These thin labels are suitable for use with microscopy slides.
- Permanent adhesive
- No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Specifiche
- Colore:nero su bianco
- Descrizione:Slide labels on sheets, custom imprint, tag
- Dimensioni:22×22 mm
- Materiale:Paper
- Seq:005