Microscope slide boxes
50 slides, nero/bianco, Legno, 50
Codice catalogo 631-9012
Nome del fornitore: BERGMANN KARTON
Vai a
Product Details & Documents
These boxes are designed for 25, 50 and 100 slides.
- Metal hinges and snap closure
- Numbered racks with table of contents
- Solid design with wooden frame and black coating
Specifiche
- N. di vetrini:50
- Colore:nero/bianco
- Descrizione:50 slides
- Materiale:Legno
- Cat. No.:631-9012