Binocular magnifiers
Binocular magnifying lens with 130 mm working distance, 3×
Catalog # 630-8216
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
Magnifiers with discrete metal frame and black matt finish for use with monocular lens components.
* Shorter working distance due to integrated convergence supporting prisms.
Delivery information: Supplied with a protective box.
Caution: Not suitable for spectacle wearers.
Specifications
- Ingrandimento:3×
- Descrizione:Binocular magnifying lens with 130 mm working distance
- Seq:003