Orders LinkContactUsLinkComponent
Standard microliter syringes
Standard microliter syringes
250 µl, 1
Codice catalogo 613-1719
Nome del fornitore:   Hospidex
Standard microliter syringes
Codice catalogo 613-1719
Nome del fornitore:  Hospidex
Standard microliter syringes
Standard microliter syringes
250 µl, 1
Price
105.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Providing ultra-precise analyses. The plunger extends down into the tip of the needle ensuring that no dead volume is created.
The bevelled needle tip is 50 mm long.
Specifiche
  • Tipo punta:
    1
  • Volume:
    250 µl